Key Documents
BR122564
BRAND® laboratory bottle, graduated with screw cap
capacity 2000 mL
Synonym(s):
Bottles, Glass, Laboratory bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
autoclavable
graduations
with cap
packaging
pack of 10 ea
manufacturer/tradename
BRAND 122564
capacity
2000 mL
diam. × H
136 mm × 268 mm
General description
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)