ALD00364
4-(2-Bromoacetyl)benzenesulfonyl fluoride
95% (GC)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H6BrFO3S
Molecular Weight:
281.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95% (GC)
form
crystals
reaction suitability
reaction type: click chemistry
mp
104-110 °C
density
1.705 g/cm3 at 20 °C
SMILES string
FS(C1=CC=C(C(CBr)=O)C=C1)(=O)=O
InChI
1S/C8H6BrFO3S/c9-5-8(11)6-1-3-7(4-2-6)14(10,12)13/h1-4H,5H2
InChI key
RWHNIEOAPNZZGI-UHFFFAOYSA-N
Related Categories
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service