A50401
2-Amino-4,6-dihydroxypyrimidine
98%
Synonym(s):
2-Amino-4,6-pyrimidinediol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
>300 °C (lit.)
SMILES string
Nc1nc(O)cc(O)n1
InChI
1S/C4H5N3O2/c5-4-6-2(8)1-3(9)7-4/h1H,(H4,5,6,7,8,9)
InChI key
AUFJTVGCSJNQIF-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kollur Shiva Prasad et al.
Sensors (Basel, Switzerland), 18(8) (2018-08-18)
In the present study, we describe the facile synthesis of silver nanoparticles (AgNPs) and their nanostructures functionalized with 2-aminopyrimidine-4,6-diol (APD-AgNPs) for Hg2+ ion detection. The promising colorimetric response of APD-AgNPs to detect Hg2+ ions was visible with naked eyes and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service