Skip to Content
Merck
All Photos(2)

Key Documents

938688

Sigma-Aldrich

ImPy-Dipp-IMes

greener alternative

Synonym(s):

5-(2,6-diisopropylphenyl)-2-mesitylimidazo[1,5-a]pyridin-2-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33ClN2
CAS Number:
Molecular Weight:
433.03
UNSPSC Code:
12352100
NACRES:
NA.21

form

powder

Quality Level

reaction suitability

reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white

greener alternative category

SMILES string

CC1=CC(C)=CC(C)=C1[N+]2=CN3C(C4=C(C(C)C)C=CC=C4C(C)C)=CC=CC3=C2.[Cl-]

InChI

1S/C28H33N2.ClH/c1-18(2)24-11-9-12-25(19(3)4)27(24)26-13-8-10-23-16-29(17-30(23)26)28-21(6)14-20(5)15-22(28)7;/h8-19H,1-7H3;1H/q+1;/p-1

InChI key

BQLNHZQDABCIAO-UHFFFAOYSA-M

General description

ImPy-Dipp-IMes is an NHC precursor developed in the Szostak group. The ligand is useful in a range of catalytic transformations, and is particularly well suited to the following: Suzuki-Miyaura cross-coupling of aryl mesylates and tosylates, Suzuki–Miyaura cross-coupling of nitroarenes (Ar–NO2), Buchwald-Hartwig amination of alkyl sulfoxides.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Suzuki-Miyaura cross-coupling of aryl mesylates and tosylates Suzuki–Miyaura cross-coupling of nitroarenes (Ar–NO2) or aryl sulfamates Buchwald-Hartwig amination of alkyl sulfoxides or aryl sulfamates.

Features and Benefits

ImPy-Dipp-Imes is an asymmetric NHC ligand precursor. It forms air-and moisture stable complexes with transition metals and is particularly effective in palladium catalyzed cross-coupling reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service