Skip to Content
Merck
All Photos(1)

Documents

727512

Sigma-Aldrich

8-(Bromomethyl)quinoline

≥96.5% (GC)

Synonym(s):

8-Quinolinylmethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BrN
CAS Number:
Molecular Weight:
222.08
Beilstein:
119140
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.5% (GC)

form

powder

SMILES string

BrCc1cccc2cccnc12

InChI

1S/C10H8BrN/c11-7-9-4-1-3-8-5-2-6-12-10(8)9/h1-6H,7H2

InChI key

IAAUGSYGHOFWEW-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kuppusamy Kanagaraj et al.
iScience, 23(3), 100927-100927 (2020-03-15)
The design and development of specific recognition and sensing systems for biologically important anionic species has received growing attention in recent years, as they play significant roles in biology, pharmacy, and environmental sciences. Herein, a new supramolecular sensing probe L1
Yi-Shan Li et al.
Talanta, 218, 121143-121143 (2020-08-17)
Dipeptidyl peptidase 4 (DPP-4) inhibitors are incretin-based medications used as oral antidiabetic agents for the treatment of type 2 diabetes. However, DPP-4 inhibitors produce side effects like acute pancreatitis, upper respiratory tract infection, nasopharyngitis, urinary tract infection, serious allergies, cardiovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service