Skip to Content
Merck
All Photos(1)

Key Documents

663336

Sigma-Aldrich

(1R,2R)-(−)-trans-1-Amino-2-indanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]/D -23.0°, c = 1 in ethanol

mp

142-146 °C

functional group

hydroxyl

SMILES string

N[C@H]1[C@H](O)Cc2ccccc12

InChI

1S/C9H11NO/c10-9-7-4-2-1-3-6(7)5-8(9)11/h1-4,8-9,11H,5,10H2/t8-,9-/m1/s1

InChI key

LOPKSXMQWBYUOI-RKDXNWHRSA-N

Application

(1R,2R)-(−)-trans-1-Amino-2-indanol has been used as a starting material to prepare an indene-based chiral auxiliary, which is used in the aldol reaction.
It can also be used as:
  • A starting material in the synthesis of oxazoline-alcohol ligands, which are employed in the asymmetric addition reaction of diethylzinc to aldehydes.
  • A chiral test compound in the study of enantiomeric separation of chiral primary amines using supercritical fluid chromatography (SFC) and HPLC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of enantiomeric separations and screening protocols for chiral primary amines by SFC and HPLC
Armstrong DW, et al.
LCGC North America, 32(17), 742-752 (2014)
Efficient one-step synthesis of chiral bidentate oxazoline-alcohol ligands via a cyclic imidate ester rearrangement
Noel T, et al.
Tetrahedron Asymmetry, 20(17), 1962-1968 (2009)
Indene-based thiazolidinethione chiral auxiliary for propionate and acetate aldol additions
Osorio-Lozada A and Olivo HF
Organic Letters, 10(4), 617-620 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service