631396
4-(Aminomethyl)benzonitrile hydrochloride
97%
Synonym(s):
4-Cyanobenzylamine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NH2CH2C6H4CN · HCl
CAS Number:
Molecular Weight:
168.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
274-279 °C (lit.)
functional group
amine
nitrile
SMILES string
Cl[H].NCc1ccc(cc1)C#N
InChI
1S/C8H8N2.ClH/c9-5-7-1-2-8(6-10)4-3-7;/h1-4H,5,9H2;1H
InChI key
QREZLLYPLRPULF-UHFFFAOYSA-N
Related Categories
Application
4-(Aminomethyl)benzonitrile hydrochloride can be used in the synthesis of fluorescent-labeled bisbenzamidine, which can be a biochemical tool in biomedical studies. It can also be used in the synthesis of 3-(p-benzylamino)-1,2,4,5 tetrazine, used in the modification of alginate hydrogels, employed in cell engineering applications.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
"Versatile click alginate hydrogels crosslinked via tetrazine?norbornene chemistry"
Desai.MR, et al.
Biomaterials, 50, 30-37 (2015)
"Synthesis of a Fluorescent-Labeled Bisbenzamidine Containing the Central (6, 7-Dimethoxy-4-coumaryl) Alanine Building Block"
Haubler D and Gutschow M
Heteroatom Chem., 26(05), 367-337 (2015)
R Voegeli et al.
International journal of cosmetic science, 39(2), 109-120 (2016-07-20)
The aim of this study was to optimize the synthesis of the plasmin and urokinase (uPA) inhibitor benzylsulfonyl-D-Ser-homoPhe-(4-amidino-benzylamide) (BSFAB), to characterize its activity and mechanism of action and to assess its use to improve stratum corneum (SC) barrier function. Peptide
Daru Seto Bagus Anugrah et al.
Carbohydrate polymers, 223, 115070-115070 (2019-08-21)
A biodegradable, near-infrared (NIR) - responsive hydrogel is one of the most promising strategies as a remotely triggered drug carrier. In this study, novel NIR-responsive hydrogels based on alginate structures were prepared for controllable drug release. The hydrogels were formed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service