Skip to Content
Merck
All Photos(1)

Documents

616486

Sigma-Aldrich

2,4-Dinitrotoluene-3,5,6-d3

98 atom % D

Synonym(s):

6-methyl-3,5-dinitro-benzene-1,2,4-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D3H3N2O4
CAS Number:
Molecular Weight:
185.15
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mp

67-70 °C (lit.)

mass shift

M+3

SMILES string

[2H]c1c([2H])c(c([2H])c(c1C)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3/i2D,3D,4D

InChI key

RMBFBMJGBANMMK-NRUYWUNFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service