Skip to Content
Merck
All Photos(1)

Documents

545090

Sigma-Aldrich

Europium(III) acetate hydrate

99.999% trace metals basis

Synonym(s):

Europium acetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
329.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

core: europium
reagent type: catalyst

SMILES string

O.CC(=O)O[Eu](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Eu.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

JGJIXMFWDPQYRL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service