Skip to Content
Merck
All Photos(1)

Documents

528382

Sigma-Aldrich

2-Methoxy-1-naphthoic acid

98%

Synonym(s):

2-Methoxynaphthoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CO2H
CAS Number:
Molecular Weight:
202.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

177-180 °C (lit.)

SMILES string

COc1ccc2ccccc2c1C(O)=O

InChI

1S/C12H10O3/c1-15-10-7-6-8-4-2-3-5-9(8)11(10)12(13)14/h2-7H,1H3,(H,13,14)

InChI key

OSTYZAHQVPMQHI-UHFFFAOYSA-N

General description

2-Methoxy-1-naphthoic acid is the 3-methoxy derivative of 1-napthoic acid. It can be prepared from a Grignard reagent derived from 1-bromo-2-methoxynaphthalene. 2-Methoxy-1-naphthoic acid undergoes Birch reduction to afford 2-methoxy-1,4,5,8-tetrahydro-1-naphthoic acid.

Application

2-Methoxy-1-naphthoic acid may be used in the synthesis of chiral (methoxynaphthyl)oxazoline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of binaphthyls via nucleophilic aromatic substitution on chiral oxazolines.
Meyers AI and Lutomski KA.
Journal of the American Chemical Society, 104(3), 879-881 (1982)
Birch Reduction of 2-Naphthoic and of ortho-Methoxynaphthoic Acids.
Eliel EL and Hoover TE.
The Journal of Organic Chemistry, 24(7), 938-942 (1959)
The Reaction of Phenyl 2-Methoxy-1-naphthoate with Grignard Reagents. A New Route to Fluorenones.
Fuson RC and Wassmundt FW.
Journal of the American Chemical Society, 78(20), 5409-5413 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service