Skip to Content
Merck
All Photos(2)

Documents

526851

Sigma-Aldrich

PSS-Octaphenyl substituted

Synonym(s):

1,3,5,7,9,11,13,15-Octaphenylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Octaphenyl-POSS®, Octaphenylsilsesquioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H40O12Si8
CAS Number:
Molecular Weight:
1033.51
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mp

>350 °C (lit.)

Quality Level

SMILES string

O1[Si]2(O[Si]3(O[Si]4(O[Si]1(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)c6ccccc6)c7ccccc7)c8ccccc8)c9ccccc9)c%10ccccc%10)c%11ccccc%11)c%12ccccc%12)c%13ccccc%13

InChI

1S/C48H40O12Si8/c1-9-25-41(26-10-1)61-49-62(42-27-11-2-12-28-42)52-65(45-33-17-5-18-34-45)54-63(50-61,43-29-13-3-14-30-43)56-67(47-37-21-7-22-38-47)57-64(51-61,44-31-15-4-16-32-44)55-66(53-62,46-35-19-6-20-36-46)59-68(58-65,60-67)48-39-23-8-24-40-48/h1-40H

InChI key

KBXJHRABGYYAFC-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service