Skip to Content
Merck
All Photos(1)

Key Documents

486736

Sigma-Aldrich

Iodoethane-1,1-d2

≥98 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Ethyl-1,1-d2 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CD2I
CAS Number:
Molecular Weight:
157.98
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.509 (lit.)

bp

69-73 °C (lit.)

mp

-108 °C (lit.)

density

1.974 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

[2H]C([2H])(C)I

InChI

1S/C2H5I/c1-2-3/h2H2,1H3/i2D2

InChI key

HVTICUPFWKNHNG-CBTSVUPCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service