Skip to Content
Merck
All Photos(1)

Key Documents

463450

Sigma-Aldrich

Methyl 4-acetamido-5-chloro-2-methoxybenzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H2(Cl)(OCH3)CO2CH3
CAS Number:
Molecular Weight:
257.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

153-156 °C (lit.)

SMILES string

COC(=O)c1cc(Cl)c(NC(C)=O)cc1OC

InChI

1S/C11H12ClNO4/c1-6(14)13-9-5-10(16-2)7(4-8(9)12)11(15)17-3/h4-5H,1-3H3,(H,13,14)

InChI key

OUEXNQRVYGYGIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruben De Pauw et al.
Journal of chromatography. A, 1420, 129-134 (2015-10-20)
A new system is proposed for applying fast temperature gradients in liquid chromatography. It consists of a 0.7 mm × 150 mm fused-silica column coated with a 50 μm Nickel-layer, which is connecting with a power source and a temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service