All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6N4S
CAS Number:
Molecular Weight:
178.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
308-313 °C (lit.)
solubility
pyridine: soluble 10%, clear
SMILES string
Sc1n[nH]c(n1)-c2ccncc2
InChI
1S/C7H6N4S/c12-7-9-6(10-11-7)5-1-3-8-4-2-5/h1-4H,(H2,9,10,11,12)
InChI key
PBTAXYKMCKLQSM-UHFFFAOYSA-N
Related Categories
General description
5-(4-Pyridyl)-1H-1,2,4-triazole-3-thiol, a mercapto-substituted 1,2,4-triazole, is a versatile heterocyclic ligand having thiol group.
Application
5-(4-Pyridyl)-1H-1,2,4-triazole-3-thiol (PTT) may be used as ligand in the preparation of Zn(II) coordination frameworks under solvothermal conditions. It may be used for the encapsulation of ZnO nanorods for their surface modification.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Surface Modification of Zinc Oxide Nanorods with Zn-Porphyrin via Metal-Ligand Coordination for Photovoltaic Applications.
Koo J, et al.
Bull. Korean Chem. Soc., 33(2), 637-637 (2012)
Xu-Dong Chen et al.
Chemical communications (Cambridge, England), (11)(11), 1296-1298 (2008-04-05)
Controlled assembly of Zn(II) with a versatile ligand 5-(4-pyridyl)-1H-1,2,4-triazole-3-thiol (Hptt) in conventional or solvothermal condition affords two distinct coordination frameworks, in which the former shows a unique 3-D pseudo-polyrotaxane architecture with 1-D water tapes penetrating the 2-D Zn-ptt layers, whereas
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service