Skip to Content
Merck
All Photos(1)

Documents

295175

Sigma-Aldrich

Chlorotrifluoromethane

≥99%

Synonym(s):

CFC-13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CClF3
CAS Number:
Molecular Weight:
104.46
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥99%

SMILES string

FC(F)(F)Cl

InChI

1S/CClF3/c2-1(3,4)5

InChI key

AFYPFACVUDMOHA-UHFFFAOYSA-N

Packaging

Packaged in lecture bottle

Recommended products

Brass control valve Z146943 or either of brass regulators Z146706 and Z146714 is recommended.

Pictograms

Gas cylinderExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ozone 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service