Skip to Content
Merck
All Photos(1)

Key Documents

294934

Sigma-Aldrich

Dipropyl carbonate

99%

Synonym(s):

Di-n-propyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2O)2CO
CAS Number:
Molecular Weight:
146.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.401 (lit.)

bp

167-168 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)OCCC

InChI

1S/C7H14O3/c1-3-5-9-7(8)10-6-4-2/h3-6H2,1-2H3

InChI key

VUPKGFBOKBGHFZ-UHFFFAOYSA-N

General description

Thermal decomposition of dipropyl carbonate at 300-400°C yields carbon dioxide, alkene and alcohol. Dipropyl carbonate reacts with hydrous titanium dioxide (TiO2 ·nH2O, n=0.15-1.23) at 453-573K to yield titanium tetraalkoxides [Ti(OPrn)4]. N435-catalyzed transesterification of diethyl carbonate with dipropyl carbonate to form ethyl propyl carbonate has been reported.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of titanium tetraalkoxides from hydrous titanium dioxideand dialkyl carbonates.
Suzuki E, et al.
Journal of Materials Chemistry, 7(10), 2049-2051 (1997)
Controlled lipase-catalyzed synthesis of poly (hexamethylene carbonate).
Jiang Z, et al.
Macromolecules, 40(22), 7934-7943 (2007)
The thermal decomposition of simple carbonate esters.
Cross JTD, et al.
Australian Journal of Chemistry, 29(7), 1477-1481 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service