Skip to Content
Merck
All Photos(2)

Key Documents

279528

Sigma-Aldrich

1,3-Diiminoisoindoline

97%

Synonym(s):

1,3-Isoindolinediimine, Phthalimide diimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3
CAS Number:
Molecular Weight:
145.16
Beilstein:
118477
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder or crystals

mp

~197 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N=C1NC(=N)c2ccccc12

InChI

1S/C8H7N3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H,(H3,9,10,11)

InChI key

RZVCEPSDYHAHLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey M Borisov et al.
ACS applied materials & interfaces, 2(2), 366-374 (2010-02-27)
A new class of oxygen indicators is described. Platinum(II) and palladium(II) complexes of azatetrabenzoporphyrins occupy an intermediate position between tetrabenzoporphyrins and phthalocyanines and combine features of both. The new dyes are excitable in the red part of the spectrum and
Nathan J Yutronkie et al.
Materials (Basel, Switzerland), 12(8) (2019-04-27)
Efficient synthesis of silicon phthalocyanines (SiPc) eliminating the strenuous reaction conditions and hazardous reagents required by classical methods is described. Implementation into organic thin-film transistors (OTFTs) affords average electron field-effect mobility of 3.1 × 10-3 cm2 V-1 s-1 and threshold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service