Skip to Content
Merck
All Photos(1)

Key Documents

272361

Sigma-Aldrich

2,3-Diaminotoluene

97%

Synonym(s):

3-Methyl-o-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein:
907184
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

59-65 °C (lit.)

SMILES string

Cc1cccc(N)c1N

InChI

1S/C7H10N2/c1-5-3-2-4-6(8)7(5)9/h2-4H,8-9H2,1H3

InChI key

AXNUJYHFQHQZBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The antitumour activity of Pd(II) and Pt(II) new complexes with 2,3-diaminotoluene was studied. 2,3-Diaminotoluene induced CYP1A activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pérez-Cabré et al.
Journal of inorganic biochemistry, 98(3), 510-521 (2004-02-28)
Pd(II) and Pt(II) new complexes with simple aromatic diamines were synthesised and characterised with the aim of studying their possible antitumour activity. The aromatic diamines chosen were 2,3-diaminotoluene (2,3 dat), 3,4-diaminotoluene (3,4 dat), 4,5-diaminoxylene (4,5 dax) and 2,3-diaminophenol (2,3 dap).
Y L Cheung et al.
Toxicology and applied pharmacology, 139(1), 203-211 (1996-07-01)
The present study was undertaken to provide a rationale for the marked difference in carcinogenic potential among isomeric diaminotoluenes, in relation to their ability to induce their own bioactivation through CYP1A induction, their genotoxic potential, and their ability to bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service