Skip to Content
Merck
All Photos(2)

Key Documents

259179

Sigma-Aldrich

3,4-Dichlorobenzyl chloride

97%

Synonym(s):

α,3,4-Trichlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2Cl
CAS Number:
Molecular Weight:
195.47
Beilstein:
386644
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.577 (lit.)

bp

122-124 °C/14 mmHg (lit.)

density

1.411 g/mL at 25 °C (lit.)

SMILES string

ClCc1ccc(Cl)c(Cl)c1

InChI

1S/C7H5Cl3/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,4H2

InChI key

YZIFVWOCPGPNHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dichlorobenzyl chloride has been used:
  • in Friedel-Crafts synthesis of 4-(3,4-dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, a key intermediate for the synthesis of sertraline
  • as alkylating agent in synthesis of poly(ether ketone)s having pendant sulfonic acid groups
  • in preparation of 5,6-dichloro-2-(3,4-dichlorobenzylthio)benzimidazole

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedel-Crafts synthesis of 4-(3, 4-dichlorophenyl)-3, 4-dihydro-1 (2H)-naphthalenone, a key intermediate in the preparation of the antidepressant sertraline.
Quallich GJ, et al.
The Journal of Organic Chemistry, 55(16), 4971-4973 (1990)
Syntheses of branched poly (ether ketone) s with pendant functional groups based on 1, 1, 1-tris (4-hydroxyphenyl) ethane.
Fritsch D, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 39(11), 1335-1347 (2002)
Zygmunt Kazimierczuk et al.
Acta biochimica Polonica, 49(1), 185-195 (2002-07-26)
Two series of benzimidazole derivatives were sythesised. The first one was based on 5,6-dinitrobenzimidazole, the second one comprises 2-thioalkyl- and thioaryl-substituted modified benzimidazoles. Antibacterial and antiprotozoal activity of the newly obtained compounds was studied. Some thioalkyl derivatives showed remarkable activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service