Skip to Content
Merck
All Photos(1)

Documents

225045

Sigma-Aldrich

2-(Phenylsulfonyl)aniline

97%

Synonym(s):

2-Aminophenyl phenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2C6H4NH2
CAS Number:
Molecular Weight:
233.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

120-122 °C (lit.)

SMILES string

Nc1ccccc1S(=O)(=O)c2ccccc2

InChI

1S/C12H11NO2S/c13-11-8-4-5-9-12(11)16(14,15)10-6-2-1-3-7-10/h1-9H,13H2

InChI key

JBCUKQQIWSWEOK-UHFFFAOYSA-N

Application

2-(Phenylsulfonyl)aniline may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service