Skip to Content
Merck
All Photos(1)

Key Documents

17753

Sigma-Aldrich

2-Chlorotrityl hydrazine, polymer-bound

100-200 mesh, extent of labeling: ~0.5 mmol/g loading

Synonym(s):

(α-Hydrazino-α-phenyl-2-chlorobenzyl)polystyrene, Hydrazine bound to 2-chloro-trityl resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.5 mmol/g loading

matrix

polystyrene, crosslinked with 1% DVB

particle size

100-200 mesh

application(s)

peptide synthesis

functional group

hydrazine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Acid labile resin for the synthesis of peptide hydrazides and alkylhydrazones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Stravropulos et al.
Lett. Pept. Sci., 2, 315-315 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service