Skip to Content
Merck
All Photos(3)

Key Documents

162167

Sigma-Aldrich

3-Cyclohexene-1-methanol

98%

Synonym(s):

1,2,3,6-Tetrahydrobenzyl alcohol, Cyclohexen-4-ylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CH2OH
CAS Number:
Molecular Weight:
112.17
Beilstein:
2038546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

190-192 °C (lit.)
80-85 °C/18 mmHg (lit.)

density

0.961 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1CCC=CC1

InChI

1S/C7H12O/c8-6-7-4-2-1-3-5-7/h1-2,7-8H,3-6H2

InChI key

VEIYJWQZNGASMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Cyclohexene-1-methanol is the principal constituent of essential oil isolated from Pinus sylvestris.

Application

3-Cyclohexene-1-methanol was used in the synthesis of novel macromonomer of epoxy end-functionalized polystyrene via atom transfer radical polymerization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of cyclohexene oxide functional polystyrene macromonomers by ATRP and their use in photoinitiated cationic polymerization.
Degirmenci M, et al.
Reactive functional Polymers, 70(1), 28-34 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service