Skip to Content
Merck
All Photos(1)

Key Documents

130443

Sigma-Aldrich

2-Furoic hydrazide

98%

Synonym(s):

2-Furoic acid hydrazide, 2-Furoylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
114435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

77-79 °C (lit.)

functional group

amine
hydrazine

SMILES string

NNC(=O)c1ccco1

InChI

1S/C5H6N2O2/c6-7-5(8)4-2-1-3-9-4/h1-3H,6H2,(H,7,8)

InChI key

SKTSVWWOAIAIKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Furoic hydrazide (2-Furoylhydrazine) was used to study the Fourier transform infrared spectra, 1H NMR and 13C NMR spectra. It was also used as a reagent for determination of carbohydrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lever et al.
Analytical biochemistry, 139(1), 205-211 (1984-05-15)
4- Hydroxybenzoylhydrazine ( PAHBAH ) reacts with glucose in hot aqueous solution when alkali exceeds aroylhydrazine concentration. The related 2- furoylhydrazine ( FAH ) reacts at lower alkali concentrations, making this an attractive alternative carbohydrate reagent since it is (unlike
V Vimalraj et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 670-675 (2010-12-31)
The Fourier transform infrared spectra, 1H NMR and 13C NMR spectra of 2-furoic hydrazide have been recorded. Optimized geometry, frequency and intensity of the vibrational bands of 2-furoic hydrazide were obtained by the density functional theory (DFT) and ab initio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service