Merck
  • Oxytocin in the paraventricular nucleus attenuates incision-induced mechanical allodynia.
Oxytocin in the paraventricular nucleus attenuates incision-induced mechanical allodynia.
Experimental and therapeutic medicine (2015-03-18)
Yanfeng Zhang, Yong Yang, Ruping Dai, Hui Wu, Changqi Li, Qulian Guo
ABSTRACT
Oxytocin (OT) neurons localized in the paraventricular nucleus (PVN) and supraoptic nucleus (SON) send fibers to the brain and spinal cord. While most previous studies have looked at the role of OT in chronic pain, few have investigated the role of OT in acute pain, particularly postoperative pain. In the present study, the role of OT in incision-induced allodynia was explored for the first time, using a rat incisional pain model. Immunohistochemical staining showed that, compared with the baseline (prior to incision) measurements, the OT content in the PVN was significantly decreased at 0.5, 1.0 and 3.0 h post-incision and returned to the baseline level at 6.0 h post-incision. By contrast, there was no significant difference in the OT content in the SON prior to and subsequent to incision. A dose-dependent inhibition of mechanical hypersensitivity was detected 30 min after intracerebroventricular injection of OT (100, 400 or 600 ng) and lasted for 3.0 h. No significant difference was noted, however, between the intrathecal OT injection group (600 ng) and the control group. In conclusion, the present study provides the first
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Isoflurane, European Pharmacopoeia (EP) Reference Standard