Merck
  • Immunomodulatory effects of vitamin D on innate and adaptive immune responses to Streptococcus pneumoniae.
Immunomodulatory effects of vitamin D on innate and adaptive immune responses to Streptococcus pneumoniae.
The Journal of infectious diseases (2013-08-08)
Marie Olliver, Laura Spelmink, Jeffni Hiew, Ulf Meyer-Hoffert, Birgitta Henriques-Normark, Peter Bergman
ABSTRACT
Streptococcus pneumoniae forms part of the normal nasopharyngeal flora but can also cause a broad spectrum of inflammatory diseases. Vitamin D has potent effects on human immunity, including induction of antimicrobial peptides and suppression of T-cell proliferation, but its ability to modulate the immune response to pneumococci is unknown. Monocyte-derived dendritic cells (DCs) were stimulated with pneumococcal peptidoglycan (PGN) in the presence or absence of vitamin D. Expression of maturation markers, cytokines, pattern recognition receptors, and antimicrobial peptides were measured with flow cytometry, enzyme-linked immunosorbent assay and quantitative polymerase chain reaction. Stimulated DCs were cocultured with autologous T-helper cells, and concentrations of T-helper (Th) 1-, Th17-, and regulatory T-cell-related cytokines were measured with enzyme-linked immunosorbent assay. Vitamin D enhanced DC maturation and expression of the migration marker C-C chemokine receptor type 7 (CCR7) in PGN-stimulated cells. It also enhanced expression of key pattern recognition receptors (Toll-like receptor 2, Nucleotide-binding oligomerization domain-containing protein 2 [Nod2]) and induced a synergistic up-regulation of the inflammatory mediator IL-1β and the β-defensin Human Beta Defensin 3 (hBD-3). Furthermore, vitamin D skewed the DC-mediated T-helper response to PGN from an inflammatory Th1/Th17 phenotype toward a regulatory T-cell phenotype. Vitamin D modulates key elements of innate immunity while dampening adaptive immune responses in DCs after pneumococcal challenge, which may have implications for prevention and treatment of pneumococcus-induced inflammation.
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder