MilliporeSigma
  • Increased life span from overexpression of superoxide dismutase in Caenorhabditis elegans is not caused by decreased oxidative damage.
Increased life span from overexpression of superoxide dismutase in Caenorhabditis elegans is not caused by decreased oxidative damage.
Free radical biology & medicine (2011-08-16)
Filipe Cabreiro, Daniel Ackerman, Ryan Doonan, Caroline Araiz, Patricia Back, Diana Papp, Bart P Braeckman, David Gems
ABSTRACT
The superoxide free radical (O(2)(•-)) has been viewed as a likely major contributor to aging. If this is correct, then superoxide dismutase (SOD), which removes O(2)(•-), should contribute to longevity assurance. In Caenorhabditis elegans, overexpression (OE) of the major cytosolic Cu/Zn-SOD, sod-1, increases life span. But is this increase caused by enhanced antioxidant defense? sod-1 OE did not reduce measures of lipid oxidation or glycation and actually increased levels of protein oxidation. The effect of sod-1 OE on life span was dependent on the DAF-16/FoxO transcription factor (TF) and, partially, on the heat shock TF HSF-1. Similarly, overexpression of sod-2 (major mitochondrial Mn-SOD) resulted in life-span extension that was daf-16 dependent. sod-1 OE increased steady-state hydrogen peroxide (H(2)O(2)) levels in vivo. However, co-overexpression of catalase did not suppress the life-span extension, arguing against H(2)O(2) as a cause of longevity. sod-1 OE increased hsp-4 expression, suggesting increased endoplasmic reticulum (ER) stress. Moreover, longevity was partially suppressed by inactivation of ire-1 and xbp-1, mediators of the ER stress response. This suggests that high levels of SOD-1 protein may challenge protein-folding homeostasis, triggering a daf-16- and hsf-1-dependent stress response that extends life span. These findings imply that SOD overexpression increases C. elegans life span, not by removal of O(2)(•-), but instead by activating longevity-promoting transcription factors.
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.