Merck
  • Silver-catalyzed incorporation of carbon dioxide into o-alkynylaniline derivatives.
Silver-catalyzed incorporation of carbon dioxide into o-alkynylaniline derivatives.
Organic letters (2013-02-05)
Tomonobu Ishida, Satoshi Kikuchi, Tatsuyuki Tsubo, Tohru Yamada
ABSTRACT
Benzoxazine-2-one derivatives are important heterocycle structures because of their various pharmaceutical activities, though their synthetic methods had been limited. In some cases, toxic reagents, such as phosgene or carbon monoxide, are required. It was found that a silver catalyst successfully promoted the incorporation of CO(2) into o-alkynylanilines to afford the corresponding benzoxazine-2-ones bearing Z exo-olefin via 6-exo-dig cyclization at the activated C-C triple bond.
Sigma-Aldrich
Efavirenz, ≥98% (HPLC)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%