Merck
  • miR-21 mediates nickel nanoparticle-induced pulmonary injury and fibrosis.
miR-21 mediates nickel nanoparticle-induced pulmonary injury and fibrosis.
Nanotoxicology (2020-09-15)
Yiqun Mo, Yue Zhang, Rong Wan, Mizu Jiang, Youqiong Xu, Qunwei Zhang
ABSTRACT
We and other groups have demonstrated that exposure to nickel nanoparticles (Nano-Ni) results in severe and persistent lung inflammation and fibrosis, but the underlying mechanisms remain unclear. Here, we propose that miR-21 may play an important role in Nano-Ni-induced lung inflammation, injury, and fibrosis. Our dose- and time-response studies demonstrated that exposure of C57BL/6J (WT) mice to Nano-Ni resulted in upregulation of miR-21, proinflammatory cytokines, and profibrotic mediators. Histologically, exposure to Nano-Ni caused severe pulmonary inflammation and fibrosis. Based on the dose- and time-response studies, we chose a dose of 50 µg of Nano-Ni per mouse to compare the effects of Nano-Ni on WT with those on miR-21 KO mouse lungs. At day 3 post-exposure, Nano-Ni caused severe acute lung inflammation and injury that were reflected by increased neutrophil count, CXCL1/KC level, LDH activity, total protein concentration, MMP-2/9 protein levels and activities, and proinflammatory cytokines in the BALF or lung tissues from WT mice, which were confirmed histologically. Although Nano-Ni had similar effects on miR-21 KO mice, the above-mentioned levels were significantly lower than those in WT mice. Histologically, lungs from WT mice exposed to Nano-Ni had infiltration of a large number of polymorphonuclear (PMN) cells and macrophages in the alveolar space and interstitial tissues. However, exposure of miR-21 KO mice to Nano-Ni only caused mild acute lung inflammation and injury. At day 42 post-exposure, Nano-Ni caused extensive pulmonary fibrosis and chronic inflammation in the WT mouse lungs. However, exposure of miR-21 KO mice to Nano-Ni only caused mild lung fibrosis and chronic lung inflammation. Our results also showed that exposure to Nano-Ni caused upregulation of TGF-β1, phospho-Smad2, COL1A1, and COL3A1 in both WT and miR-21 KO mouse lungs. However, levels were significantly lower in miR-21 KO mice than in WT mice, except TGF-β1, which was similar in both kinds of mice. Decreased expression of Smad7 was observed in WT mouse lungs, but not in miR-21 KO mice. Our results demonstrated that knocking out miR-21 ameliorated Nano-Ni-induced pulmonary inflammation, injury, and fibrosis, suggesting the important role of miR-21 in Nano-Ni-induced pulmonary toxicity.
Sigma-Aldrich
Lactophenol-picric acid solution, for microscopy
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture