Skip to Content
BT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Cupped Powder Spatulas
Products
Product Category
spatulas/scoops (4)
Manufacturer
Length (cm)
Length (in)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BAF369310250
SP Bel-Art
®
Sterileware
™
Cupped Powder Spatulas,
white polystyrene, blade L × width 9.875 in. (25 cm) × 0.75 in. (18 mm), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369310150
SP Bel-Art
®
Sterileware
™
Cupped Powder Spatulas,
white polystyrene, blade L × width 15 cm (5.875 in.) × 0.75 in. (18 mm), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369320250
SP Bel-Art
®
Sterileware
™
Cupped Powder Spatulas,
blue polystyrene, blade L × width 25 cm (9.875 in.) × 0.75 in. (18 mm), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369320150
SP Bel-Art
®
Sterileware
™
Cupped Powder Spatulas,
blue polystyrene, blade L × width 5.875 in. (15 cm) × 0.75 in. (18 mm), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing