Skip to Content
Merck
All Photos(1)

Key Documents

1683708

USP

Triethylene glycol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein:
969357
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

5.2 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

autoignition temp.

699 °F

expl. lim.

9.2 %

manufacturer/tradename

USP

refractive index

n20/D 1.455 (lit.)

bp

125-127 °C/0.1 mmHg (lit.)

mp

−7 °C (lit.)

density

1.124 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCOCCOCCO

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triethylene glycol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Disclaimer

The product is notintended for use as a biocide under global biocide regulations, including butnot limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act,European Biocidal Products Regulation, Canada’s Pest Management RegulatoryAgency, Turkey’s Biocidal Products Regulation, Korea’s Consumer ChemicalProducts and Biocide Safety Management Act (K-BPR) and others.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service