Skip to Content
Merck
All Photos(1)

Key Documents

RAD302

Supelco

radiello Aldehyde Calibration Standard

50 μg/mL each component in acetonitrile, pkg of 10 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NB.24

packaging

pkg of 10 mL

concentration

50 μg/mL each component in acetonitrile

application(s)

environmental

General description

The aldehyde calibration standard consists of nine 2,4-dinitrophenylhydrazones (2,4-DNPH) diluted in acetonitrile. Actual concentrations for each component are certified for each lot. The standard stock solution is shipped in a pierceable-septum crimped cap vial, and is stable for at least 4 months when stored at a 4 °C dark environment.

Note: All components are 50 μg/mL with the exception of acrolein (10 μg/mL).

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Analyte

Description

    Formaldehyde-2,4-dinitrophenylhydrazone
    Acetaldehyde-2,4-dinitrophenylhydrazone
    Acrolein-2,4-dinitrophenylhydrazone
    Hexanal 2,4-dinitrophenylhydrazone
    Benzaldehyde-2,4-dinitrophenylhydrazone

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile aldehydes in libraries and archives
Fenech, A., et al.
Atmospheric Environment, 44 (17), 2067-2073 (2010)
Ambient levels and temporal trends of VOCs, including carbonyl compounds, and ozone at Caba?eros National Park border, Spain
Villanueva, F., et al.
Atmospheric Environment, 85, 256-265 (2014)
Evaluation of a diffusive sampler for measurement of carbonyl compounds in air
Uchiyama, S., et al.
Atmospheric Environment, 38 (37), 6319-6326 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service