PHL86708
Lupulone (stable DCHA salt)
phyproof® Reference Substance
Synonym(s):
Lupulone (dicyclohexylammonium) salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H38O4·C12H23N
CAS Number:
Molecular Weight:
595.90
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
pharmaceutical
storage temp.
−20°C
Related Categories
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Analysis Note
The counterion is determined quantitatively and treated as an impurity in the calculation of the absolute purity of this reference substance. The assigned absolute purity on the certificate of analysis of this reference standard, therefore, refers to the pure lupulone.
Other Notes
This compound is commonly found in plants of the genus: humulus
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service