Skip to Content
Merck
All Photos(1)

Key Documents

47537-U

Supelco

52 Component Indoor Air Standard

certified reference material, 100 μg/mL each component in methanol: water (95:5)

Synonym(s):

Japanese Indoor Air Standard Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

100 μg/mL each component in methanol: water (95:5)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetone
    Benzene
    Bromodichloromethane
    1-Butanol
    2-Butanone
    Butyl acetate
    Carbon tetrachloride
    Chloroform
    Decanal
    Decane
    Dibromochloromethane
    1,4-Dichlorobenzene
    1,2-Dichloroethane
    Dichloromethane
    1,2-Dichloropropane
    2,4-Dimethylpentane
    Dodecane
    Ethanol
    Ethyl acetate
    Ethylbenzene
    2-Ethyltoluene
    3-Ethyltoluene
    4-Ethyltoluene
    Heptane
    Hexadecane
    Hexane
    (R)-(+)-Limonene
    4-Methyl-2-pentanone
    Nonanal
    Nonane
    Octane
    Pentadecane
    (−)-α-Pinene
    (−)-β-Pinene
    1-Propanol
    2-Propanol
    Styrene
    Tetrachloroethylene
    Tetradecane
    Durene
    Toluene
    1,1,1-Trichloroethane
    Trichloroethylene
    Tridecane
    1,2,3-Trimethylbenzene
    1,2,4-Trimethylbenzene
    Mesitylene
    2,2,4-Trimethylpentane
    Undecane
    m-Xylene
    o-Xylene
    p-Xylene
    See All (52)

Signal Word

Danger

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service