Skip to Content
Merck
All Photos(2)

Documents

29755-U

Supelco

Graphsphere 2016

greener alternative

glass TD tube, O.D. × L 1/4 × 3 1/2, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

glass TD tube

Quality Level

description

For use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments, PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

feature

fritted
preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

O.D. × L

1/4 × 3 1/2

matrix

Carboxen® 1016 carbon molecular sieve (CMS)

suitability

suitable for (An Alternative to Tenax TA)

application(s)

environmental
industrial hygiene

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
Graphsphere is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service