22273
Valco® Precision Sampling Syringe, Series A-2, Removable needle
volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
Synonym(s):
SYRINGE,PS-A-2,1ML
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
bevel tip stainless steel needle (point style 2)
description
Series A-2
feature
removable needle Series A2
packaging
pkg of 1 ea
manufacturer/tradename
VICI 050033
needle L × O.D. × I.D.
2 in. × 0.029 in. × 0.012 in.
volume
1000 μL
Related Categories
General description
Series A-2 - Removable Needle: An economically priced general purpose syringe, it is leak-tight to 250 psi for both gas and liquids.
- Push-button valve - allows instantaneous injection
- Smaller volumes - great for small volatile samples
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)