Skip to Content
Merck
All Photos(1)

Key Documents

Z617911

Greiner centrifuge tubes with screw cap

capacity 12 mL, size × diam. × L × 16.8 mm × 100 mm, conical bottom, skirt

Synonym(s):

centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, screw cap centrifuge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

colorless polystyrene tube
conical bottom

sterility

non-sterile

feature

RCF 5,000 × g
skirt

packaging

case of 1300

manufacturer/tradename

Greiner 164180

capacity

12 mL

size diam. × L

× 16.8 mm × 100 mm

color

white cap, polyethylene

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene tubes with closures are ideally suited for optical measurements due to its high clarity. Available with screw or bayonetc ap in blue, black or white and choice of skirted on non-skirted.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service