VIV405001
SynVivo SynALI 3D Air Liquid Interface Lung model starter kit, includes pneumatic priming device
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
Recommended Products
packaging
case of 1 ea
manufacturer/tradename
SynVivo 405001
storage temp.
room temp
General description
10 - IMN2-LC 3 μm slit barrier chips, 100 ft Tygon Tubing, 25 pack - Slide Clamps, 50 pack - 1 mL Syringe with Luer-Lok Tip, 50 pack - 24 Gauge Needle x 0.5 in long, SynVivo Pneumatic Primer, 2 - Five Stopcock Manifold 4-Way all FLL
SynVivo has developed a novel Air Liquid Interface (ALI) model mimicking the lung architecture. The device comprises of a plastic, disposable and optically clear microfluidic chip functionalized with epithelial cells surrounded by vasculature comprised of endothelial cells. This structure maintains an Air Liquid Interface across the airway cells, allowing the formation of airways tubules that transport mucus and are maintained by the surrounding epithelium. Cell morphology, airway structure, cell-cell interactions and functions of the airway (e.g. mucus transport, ciliary beating, therapeutic-induced improvements) can be visualized and quantified in real-time in both normal and diseased conditions.
Unique Features include:
SynVivo has developed a novel Air Liquid Interface (ALI) model mimicking the lung architecture. The device comprises of a plastic, disposable and optically clear microfluidic chip functionalized with epithelial cells surrounded by vasculature comprised of endothelial cells. This structure maintains an Air Liquid Interface across the airway cells, allowing the formation of airways tubules that transport mucus and are maintained by the surrounding epithelium. Cell morphology, airway structure, cell-cell interactions and functions of the airway (e.g. mucus transport, ciliary beating, therapeutic-induced improvements) can be visualized and quantified in real-time in both normal and diseased conditions.
Unique Features include:
- Morphologically realistic airway structure and environment
- Air Liquid Interface (ALI) across the epithelium and endothelium
- Real-time visualization of cellular and barrier functionality including mucus, ciliary beating, immune cell interactions and therapeutic screening
- Robust and easy to use protocols
- Available as chips and starter kits or for services based screening
Linkage
LInk to Publications
application text
application text
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service