Key Documents
TMO312085-0004
Nalgene® narrow-mouth bottle
amber high-density polyethylene, volume 125 mL, case of 500 ea
Synonym(s):
Nalgene bottle, plastic bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
amber high-density polyethylene
packaging
case of 500 ea
manufacturer/tradename
Nalgene 312085-0004
capacity
125 mL
volume
125 mL
General description
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)