Skip to Content
Merck
All Photos(1)

Key Documents

SRP0412

Sigma-Aldrich

Sestrin 2 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥35% (SDS-PAGE)

Synonym(s):

HI95, SES2, SEST2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥35% (SDS-PAGE)

form

aqueous solution

mol wt

55 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SESN2(83667)

General description

Sestrin 2 (SESN2), a highly conserved protein, is a member of a family of anti-aging proteins called the sestrins. Human SESN2 (hSESN2) contains three subdomains, such as SESN-A, SESN-B, and SESN-C. The SESN2 gene is mapped to human chromosome 1p35.3.

Application

Sestrin 2 human may be used to study its role on blood brain- barrier (BBB) stabilization after moderate and severe hypoxic- ischemic (HI) injuries in neonatal rats.

Biochem/physiol Actions

Sestrin 2 (SESN2) plays a vital role in modulating cellular stress response and oxidant defense. It plays a regulatory role in energy metabolism, cell fate, liver diseases, inflammation, and carcinogenesis. This novel anti-aging protein aids to maintain mitochondrial activities and enhance mitochondrial health. Since SESN2 is capable of protecting tissues from oxidative stress and mitochondrial dysfunction caused by ischemia or reperfusion (I/R), it may be a promising therapeutic target in hepatic regeneration.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raúl P Oliveira et al.
Free radical biology & medicine, 163, 255-267 (2020-12-29)
Liver regeneration is a remarkably complex phenomenon conserved across all vertebrates, enabling the restoration of lost liver mass in a matter of days. Unfortunately, extensive damage to the liver may compromise this process, often leading to the death of affected
Li-Xue Wang et al.
Frontiers in immunology, 10, 2797-2797 (2019-12-24)
Sestrin2 (SESN2), a highly evolutionarily conserved protein, is critically involved in cellular responses to various stresses. SESN2 has a protective effect on physiological and pathological states mainly via regulating oxidative stress, endoplasmic reticulum stress, autophagy, metabolism, and inflammation. In recent
Xudan Shi et al.
Neurobiology of disease, 95, 111-121 (2016-07-19)
Hypoxic ischemic (HI) encephalopathy remains the leading cause of perinatal brain injury resulting in long term disabilities. Stabilization of blood brain barrier (BBB) after HI is an important target, therefore, in this study we aim to determine the role of
Dongdong Wu et al.
Life sciences, 267, 118941-118941 (2020-12-29)
Acute lung injury (ALI) / acute respiratory distress syndrome (ARDS) is a critical clinical syndrome with complex pathology and pathogenesis. Since there is no specific treatment for ALI, it is important to study the mechanism of how ALI develop. Sestrin2
Regine Bergholdt et al.
PloS one, 4(7), e6250-e6250 (2009-07-18)
Proteins contributing to a complex disease are often members of the same functional pathways. Elucidation of such pathways may provide increased knowledge about functional mechanisms underlying disease. By combining genetic interactions in Type 1 Diabetes (T1D) with protein interaction data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service