Skip to Content
Merck
All Photos(1)

Key Documents

SML3276

Sigma-Aldrich

Apelin-17 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Apelin 17 trifluoroacetate salt, K17F trifluoroacetate salt, L-Lysyl-L-phenylalanyl-L-arginyl-L-arginyl-L-glutaminyl-L-arginyl-L-prolyl-L-arginyl-L-leucyl-L-seryl-L-histidyl-L-lysylglycyl-L-prolyl-L-methionyl-L-prolyl-L-phenylalanine trifluoroacetate salt, Lys-Phe-Arg-Arg-Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Assay:
≥95% (HPLC)
form:
powder

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

Apelin-17 (K17F) is a potent and selective endogenous agonist of apelin receptor (apelin-R). Apelin-17 has 16 times higher affinity for apelin-R than apelin-13 (pE13F) (Ki = 0.05 versus 0.8 nM).
potent and selective endogenous agonist of apelin receptor (apelin-R)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang Wang et al.
Hypertension (Dallas, Tex. : 1979), 68(2), 365-377 (2016-05-25)
Apelin peptides mediate beneficial effects on the cardiovascular system and are being targeted as potential new drugs. However, apelin peptides have extremely short biological half-lives, and improved understanding of apelin peptide metabolism may lead to the discovery of biologically stable
Adrien Flahault et al.
Nature communications, 12(1), 305-305 (2021-01-14)
Apelin and arginine-vasopressin (AVP) are conversely regulated by osmotic stimuli. We therefore hypothesized that activating the apelin receptor (apelin-R) with LIT01-196, a metabolically stable apelin-17 analog, may be beneficial for treating the Syndrome of Inappropriate Antidiuresis, in which AVP hypersecretion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service