Skip to Content
Merck
All Photos(1)

Documents

SML1929

Sigma-Aldrich

Equisetin

(Fusarium equiseti), ≥97% (HPLC)

Synonym(s):

(3E,5S)-3-[[(1S,2R,4aS,6R,8aR)-1,6-dimethyl-2-[(E)-prop-1-enyl]-4a,5,6,7,8,8a-hexahydro-2H-naphthalen-1-yl]-hydroxymethylidene]-5-(hydroxymethyl)-1-methylpyrrolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO4
CAS Number:
Molecular Weight:
373.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

biological source

(Fusarium equiseti)

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: soluble 1 mg/mL
chloroform: soluble 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

C[C@H]1C[C@]2([H])[C@@]([C@](C)(/C(O)=C3C([C@H](CO)N(C)C/3=O)=O)[C@H](/C=C/C)C=C2)([H])CC1

InChI

1S/C22H31NO4/c1-5-6-15-9-8-14-11-13(2)7-10-16(14)22(15,3)20(26)18-19(25)17(12-24)23(4)21(18)27/h5-6,8-9,13-17,24,27H,7,10-12H2,1-4H3/b6-5+/t13-,14-,15-,16-,17+,22-/m1/s1

InChI key

SLOQQJFOWFUUMP-KNQICWOTSA-N

Biochem/physiol Actions

Equisetin is a tetramic acid analog derived from Fusarium equisti. Equisetin is mainly known for its antibiotic and cytotoxic activity and for its inhibitory effect on HIV-1 integrase. Inhibition of HIV integrase by Equisetin was reported with IC50 values of 5-20 μM. Moreover, it was shown that Equisetin specifically inhibits the substrate anion carriers of the inner membrane of mitochondria and acts nonspecifically on protein membranes and protein hydrophobic domains. In addition, it was demonstrated that Equisetin causes phytotoxicity by causing root Necrosis and was also identified in a mycotoxin cocktail associated with cattle feeding problems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Hazuda et al.
Antiviral chemistry & chemotherapy, 10(2), 63-70 (1999-05-21)
We have identified a series of novel inhibitors of human immunodeficiency virus type 1 (HIV-1) integrase by randomly screening natural product extracts using an in vitro biochemical assay designed to identify inhibitors of integrase-catalysed strand transfer. Equisetin recovered from the
Agha W Yunus et al.
Toxins, 7(6), 2188-2197 (2015-06-16)
Cottonseed cake in South East Asia has been associated with health issues in ruminants in the recent years. The present study was carried out to investigate the health issues associated with cottonseed cake feeding in dairy animals in Pakistan. All
T König et al.
Journal of bioenergetics and biomembranes, 25(5), 537-545 (1993-10-01)
The effect of equisetin, an antibiotic produced by Fusarium equiseti, has been studied on mitochondrial functions (respiration, ATPase, ion transport). Equisetin inhibits the DNP-stimulated ATPase activity of rat liver mitochondria and mitoplasts in a concentration-dependent manner; 50% inhibition is caused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service