Skip to Content
Merck
All Photos(1)

Documents

SML0178

Sigma-Aldrich

ARL-17477 dihydrochloride hydrate

≥98% (HPLC)

Synonym(s):

N-[4-[2-[[(3-chlorophenyl)methyl]amino]ethyl]phenyl]-2-thiophenecarboximidamide dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3S·2HCl · xH2O
CAS Number:
Molecular Weight:
442.83 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

O.Cl.Cl.Clc1cccc(CNCCc2ccc(NC(=N)c3cccs3)cc2)c1

InChI

1S/C20H20ClN3S.2ClH.H2O/c21-17-4-1-3-16(13-17)14-23-11-10-15-6-8-18(9-7-15)24-20(22)19-5-2-12-25-19;;;/h1-9,12-13,23H,10-11,14H2,(H2,22,24);2*1H;1H2

InChI key

QOPATWWWZNTSQF-UHFFFAOYSA-N

Application

ARL-17477 dihydrochloride hydrate may be used in cell signaling studies.

Biochem/physiol Actions

ARL-17477 is an nNOS specific inhibitor. The IC50 against rat nNOS is 35 nM and it has 100-fold selectivity over endothelial and inducible isoforms. ARL-17477 penetrates the brain, and reduces infarct size and reduces cerebral blood flow in several different rodent models of ischemic brain injury.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

We offer many products related to nitric oxide synthases for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service