Skip to Content
Merck
All Photos(1)

Key Documents

SMB00581

Sigma-Aldrich

Narcissoside

≥95% (HPLC)

Synonym(s):

3,4′,5,7-Tetrahydroxy 3′-methoxyflavone 3-rutinoside, 3-O-Methylquercetin 3-rutinoside, 3-Rutinosylisorhamnetin, Isorhamnetin 3-O-rutinoside, Isorhamnetin 3-rhamnoglucoside, Isorhamnetol 3-rutinoside, Narcissin, Quercetin 3′-methyl ether 3-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O16
CAS Number:
Molecular Weight:
624.54
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]([C@H](CO[C@@H]1[C@@H](O)[C@@H](O)[C@H](O)[C@@H](C)O1)O2)[C@@H](O)[C@H](O)[C@H]2OC3=C(C4=CC=C(O)C(OC)=C4)OC5=CC(O)=CC(O)=C5C3=O

InChI

1S/C28H32O16/c1-9-18(32)21(35)23(37)27(41-9)40-8-16-19(33)22(36)24(38)28(43-16)44-26-20(34)17-13(31)6-11(29)7-15(17)42-25(26)10-3-4-12(30)14(5-10)39-2/h3-7,9,16,18-19,21-24,27-33,35-38H,8H2,1-2H3/t9-,16+,18-,19+,21+,22-,23+,24+,27+,28-/m1/s1

InChI key

UIDGLYUNOUKLBM-OWIBGMBDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Narcissoside is a flavonol glycoside.

Biochem/physiol Actions

Narcissoside is a phytochemical shown to have antioxidant and antiproliferative activity in cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ni et al.
Frontiers in plant science, 8, 920-920 (2017-07-01)
The flavonoids in the extracts of Ginkgo leaves have been shown to have great medical value: thus, a method to increase the flavonoid contents in these extracts is of significant importance for human health. In the present study, we investigated
Morinda citrifolia (Noni) fruit--phytochemistry, pharmacology, safety.
Potterat O and Hamburger M.
Planta Medica, 73(3), 191-199 (2007)
Parham Taslimi
Journal of biochemical and molecular toxicology, 34(11), e22566-e22566 (2020-07-03)
Tyrosinase enzyme is a functional oxidase that is extensively divided in nature. It is the main enzyme in melanin synthesis and is also involved in designating the color of mammalian hair and skin. Additionally, it is accountable for the unfavorable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service