All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C52H67N7O10
Molecular Weight:
950.13
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ac-Bhg-Leu-Asp-Ile-Ile-Trp
Application
PD-145065 is used as an endothelin-A/endothelin-B (ETA/ETB) receptor antagonist.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Comparison of 2 endothelin-receptor antagonists on in vitro responses of equine palmar digital arterial and venous rings to endothelin-1.
Stokes AM, Venugopal CS, Hosgood G, et al.
Canadian Journal of Veterinary Research = Revue Canadienne De Recherche Veterinaire, 7-, 197-205 (2006)
Paulo Castro-Chaves et al.
European journal of pharmacology, 544(1-3), 91-96 (2006-07-18)
We analyzed the influence of endothelin-1 and endocardial endothelium on the myocardial effects of angiotensin-II. Angiotensin-II (10(-9)-10(-5) M) was tested in rabbit right papillary muscles in absence (Protocol-A) or presence of PD-145065 (10(-7) M; Protocol-B), BQ-123 (10(-7) M; Protocol-C) or
Francesca Ceccarelli et al.
Biochemical pharmacology, 65(5), 783-793 (2003-03-12)
Expression and pharmacological properties of endothelin receptors (ETRs) were investigated in H9c2 cardiomyoblasts. The mechanism of receptor-mediated modulation of intracellular Ca(2+) concentration ([Ca(2+)](i)) was examined by measuring fluorescence increase of Fluo-3-loaded cells with flow cytometry. Binding assays showed that [125I]endothelin-1
K E Todd et al.
Surgery, 122(2), 443-449 (1997-08-01)
Endothelin peptides are polykines with strong vasoconstrictor properties. We have previously shown that endothelin antagonism (PD145065) reduces the local severity of acute pancreatitis. We now investigated the effect of endothelin antagonism on systemic inflammation in a model of acute hemorrhagic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service