SCP0081
Caspase 3 Apopain Substrate
≥95% (HPLC), lyophilized
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C30H34N5O13
Molecular Weight:
672.62
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
Product Name
Caspase 3 Apopain Substrate,
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥85%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ac-Asp-Glu-Val-Asp-7-AFC
Application
Ac-DEVD-7-AFC (N-acetyl-asp-glu-val-asp-7-amino-4-trifluoromethyl coumarin) is used as a cell penetrating fluorogenic CED-related caspase substrate.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Benjawan Wudtiwai et al.
Journal of hematology & oncology, 4, 52-52 (2011-12-22)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) induces apoptosis in various tumor cells, but does not affect normal cells or human leukemic cells, such as MOLT-4 and U937 cells, which are relatively resistant to TRAIL. Three flavonoids extracted from the rhizome
Kateřina Kuželová et al.
Journal of cellular biochemistry, 112(11), 3334-3342 (2011-07-14)
The effects of the pan-caspase inhibitor Q-VD-OPh on caspase activity, DNA fragmentation, PARP cleavage, 7A6 exposition, and cellular adhesivity to fibronectin were analyzed in detail in three different apoptotic systems involving two cell lines (JURL-MK1 and HL60) and two apoptosis
Caspase activation by anticancer drugs: the caspase storm.
Tao Z, Goodisman J, Penefsky HS, et al.
Molecular Pharmacology, 4, 583-595 (2007)
R A Jones et al.
Hepatology (Baltimore, Md.), 27(6), 1632-1642 (1998-06-10)
The mechanism of Fas antigen-induced hepatocyte apoptosis was investigated. Using a monoclonal antibody directed against the Fas antigen, apoptosis was induced in freshly isolated murine hepatocytes within 90 minutes of antibody addition as assessed by plasma membrane bleb formation, chromatin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service