SBCT10
SB-beta-gal-Sf9-HAM-CTRL
recombinant, expressed in baculovirus infected Sf9 cells
Synonym(s):
Negative control High Activity Memebrane (HAM) Sf9 insect membrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
form
liquid
packaging
pkg of 250 μL
concentration
5 mg/mL
shipped in
dry ice
storage temp.
−70°C
General description
General drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. Membrane Preparations for ATPase Assays are suitable for drug-efflux transporter interaction studies based on ATPase activity determination, and could be used for differentiation between transporter substrates and inhibitors. Control Membranes with no-, or significantly lower transporter activity are available for both Vesicular Transport and ATPase Assays.
Application
Control Membranes with no-, or significantly lower transporter activity provide appropriate negative controls for both Vesicular Transport and ATPase assays. Refer to individual Vesicular Transport or ATPase assay product detail pages for appropriate control membranes needed for a specific assay.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service