Skip to Content
Merck
All Photos(4)

Key Documents

SAB1400457

Sigma-Aldrich

Anti-PES1 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PES1(23481)

General description

This gene encodes a protein that is abnormally elevated in malignant tumors of astrocytic origin. It is a strongly conserved gene containing a BRCT domain that is essential for the activity of this gene product. The gene plays a crucial role in cell proliferation and may be necessary for oncogenic transformation and tumor progression. (provided by RefSeq)

Immunogen

PES1 (AAH32489.1, 1 a.a. ~ 588 a.a) full-length human protein.

Sequence
MGGLEKKKYERGSATNYITRNKARKKLQLSLADFRRLCILKGIYPHEPKHKKKVNKGSTAARTFYLIKDIRFLLHEPIVNKFREYKVFVRKLRKAYGKSEWNTVERLKDNKPNYKLDHIIKERYPTFIDALRDLDDALSMCFLFSTFPRTGKCHVQTIQLCRRLTVEFMHYIIAARALRKVFLSIKGIYYQAEVLGQPIVWITPYAFSHDHPTDVDYRVMATFTEFYTTLLGFVNFRLYQLLNLHYPPKLEGQAQAEAKAGEGTYALDSESCMEKLAALSASLARVVVPATEEEAEVDEFPTDGEMSAQEEDRRKELEAQEKHKKLFEGLKFFLNREVPREALAFIIRSFGGEVSWDKSLCIGATYDVTDSRITHQIVDRPGQQTSVIGRCYVQPQWVFDSVNARLLLPVAEYFSGVQLPPHLSPFVTEKEGDYVPPEKLKLLALQRGEDPGNLNESEEEEEEDDNNEGDGDEEGENEEEEEDAEAGSEKEEEARLAALEEQRMEGKKPRVMAGTLKLEDKQRLAQEEESEAKRLAIMMMKKREKYLYQKIMFGKRRKIREANKLAEKRKAHDEAVRSEKKAKKARPE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David P Dannheisig et al.
Cancers, 13(24) (2021-12-25)
Ribosome biogenesis is essential for protein synthesis, cell growth and survival. The process takes places in nucleoli and is orchestrated by various proteins, among them RNA polymerases I-III as well as ribosome biogenesis factors. Perturbation of ribosome biogenesis activates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service