Skip to Content
Merck
All Photos(2)

Key Documents

SAB1303578

Sigma-Aldrich

ANTI-FSTL3 (C-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

FLRG, FSTL3, Follistatin-related protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$93.10
4 X 100 ML
$322.00
1 L
$446.00

$93.10


Availability

Available to ship onDecember 23, 2024Details


To order products, please contact your local dealer.

Select a Size

Change View
100 ML
$93.10
4 X 100 ML
$322.00
1 L
$446.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$93.10


Availability

Available to ship onDecember 23, 2024Details


To order products, please contact your local dealer.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27663 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:10-1:50
western blot: 1:1000

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
D12580649621903439
Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

100

solubility

water: soluble 4.01 g/L at 20 °C

solubility

-

solubility

water: soluble 4.01 g/L at 20 °C

solubility

-

mp

<−50 °C (lit.)

mp

<−50 °C (lit.)

mp

<−50 °C (lit.)

mp

-

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n20/D 1.414 (lit.)

refractive index

n20/D 1.414 (lit.)

refractive index

n20/D 1.414 (lit.)

refractive index

n20/D 1.454

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service