Key Documents
RF4530113-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
F-45-30-11, non aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
feature
lid
to hold 30 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022654004 (NA)
Eppendorf® 5427712003 (EU)
parameter
20,817 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)