RAB1106
Rabbit IFN-gamma ELISA Kit
for cell culture supernatants, plasma, and serum samples
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rabbit
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.16 ng/ml
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rhesus macaque ... IFNG(100008602)
General description
This ELISA antibody pair detects Rabbit IFN-gamma. Other species are not determined.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service